accessibility menu, dialog, popup

UserName

CAS RN 1912-48-7

H3CNHOO

CAS RN 1912-48-7

IUPAC Name: 2-(1-methyl-1H-indol-3-yl)acetic acid
Synonyms: (1-methylindol-3-yl)acetic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: NAIPEFIYIQFVFC-UHFFFAOYSA-N
SMILES: CN1C=C(CC(O)=O)C2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results