accessibility menu, dialog, popup

UserName

CAS RN 189761-96-4

CH3NNClNOO

CAS RN 189761-96-4

IUPAC Name: 1-(2-chloro-6-nitrophenyl)-4-methylpiperazine
Synonyms: 1-(2-chloro-6-nitrophenyl)-4-methylpiperazine
Molecular Weight (g/mol): 255.7
Molecular Formula: C11H14ClN3O2
InChi Key: SKTYGQGYDUZEPT-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C(C=CC=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results