accessibility menu, dialog, popup

UserName

CAS RN 18583-60-3

BHNNNNNNK

CAS RN 18583-60-3

IUPAC Name: potassium tris(1H-pyrazol-1-yl)boranuide
Synonyms: potassium trispyrazolylborate
Molecular Weight (g/mol): 252.13
Molecular Formula: C9H10BKN6
InChi Key: IBNGZWIUPDMZMG-UHFFFAOYSA-N
SMILES: [K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)