accessibility menu, dialog, popup

UserName

CH3OOH3N

CAS RN 1836-62-0

IUPAC Name: 2-(2-methoxyphenoxy)ethan-1-aminium
Synonyms: 2-(2-methoxyphenoxy)ethanaminium
Molecular Weight (g/mol): 168.22
Molecular Formula: C9H14NO2
InChi Key: CKJRKLKVCHMWLV-UHFFFAOYSA-O
SMILES: COC1=CC=CC=C1OCC[NH3+]
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results