accessibility menu, dialog, popup

UserName

CAS RN 18162-84-0

H3CSiCH3CH3Cl

CAS RN 18162-84-0

IUPAC Name: chlorodimethyloctylsilane
Synonyms: chlorodimethyloctylsilane
Molecular Weight (g/mol): 206.83
Molecular Formula: C10H23ClSi
InChi Key: DBKNGKYVNBJWHL-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](C)(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results