accessibility menu, dialog, popup

UserName

CAS RN 1801-10-1

NH2OFFF

CAS RN 1801-10-1

IUPAC Name: 3-(trifluoromethyl)benzamide
Synonyms: 3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 189.14
Molecular Formula: C8H6F3NO
InChi Key: XBGXGCOLWCMVOI-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results