accessibility menu, dialog, popup

UserName

CAS RN 178488-36-3

IUPAC Name: 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Synonyms: 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Molecular Weight (g/mol): 220.58
Molecular Formula: C8H4ClF3N2
InChi Key: BWSHPRLSCABXRL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN2C=CN=C2C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results