accessibility menu, dialog, popup

UserName

CH3H3CClNH2

CAS RN 17797-11-4

IUPAC Name: 2-(4-chlorophenyl)propan-2-amine
Synonyms: 2-(4-chlorophenyl)propan-2-amine
Molecular Weight (g/mol): 169.65
Molecular Formula: C9H12ClN
InChi Key: JCUUNSQUTQELEZ-UHFFFAOYSA-N
SMILES: CC(C)(N)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results