CAS RN 176161-24-3
CAS RN 176161-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Maribavir, MedChemExpress
MedChemExpress Maribavir is a potent inhibitor of histone phosphorylation catalyzed by wild-type pUL97 in vitro, with an IC50 of 3 nM. Maribavir has potent antiviral activity against HCMV and Epstein-Barr virus (EBV).