accessibility menu, dialog, popup

UserName

CAS RN 175277-31-3

H3CH3CCH3SOOSH2N

CAS RN 175277-31-3

IUPAC Name: 2-(2-methylpropane-2-sulfonyl)ethanethioamide
Synonyms: 2-(2-methylpropane-2-sulfonyl)ethanethioamide
Molecular Weight (g/mol): 195.3
Molecular Formula: C6H13NO2S2
InChi Key: MNLXPMODUGFBAA-UHFFFAOYSA-N
SMILES: CC(C)(C)S(=O)(=O)CC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results