accessibility menu, dialog, popup

UserName

CAS RN 175135-93-0

CH3BrOCH3Cl

CAS RN 175135-93-0

IUPAC Name: 2-bromo-1-(3-chloro-4-methylphenyl)propan-1-one
Synonyms: 2-bromo-1-(3-chloro-4-methylphenyl)propan-1-one
Molecular Weight (g/mol): 261.54
Molecular Formula: C10H10BrClO
InChi Key: RGMFWCVOMGKIJM-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)C1=CC=C(C)C(Cl)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results