accessibility menu, dialog, popup

UserName

CAS RN 174824-16-9

IUPAC Name: 2-bromo-3,5-bis(trifluoromethyl)aniline
Synonyms: 2-bromo-3,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol): 308.02
Molecular Formula: C8H4BrF6N
InChi Key: JFWWJFXUVIYKTC-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1Br)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results