CAS RN 174574-15-3
CAS RN 174574-15-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Fluoro-N,N-dimethyl-6-nitroaniline, 97%, Thermo Scientific™
CAS: 174574-15-3 Synonym: (2-Fluoro-6-nitrophenyl)dimethylamine