accessibility menu, dialog, popup

UserName

CAS RN 17360-11-1

(S)ONH2OHHONNOOOO

CAS RN 17360-11-1

IUPAC Name: (2S)-2-amino-3-(4-hydroxy-3,5-dinitrophenyl)propanoic acid
Synonyms: 3,5-dinitrotyrosine
Molecular Weight (g/mol): 271.19
Molecular Formula: C9H9N3O7
InChi Key: SAZOSDSFLRXREA-YFKPBYRVSA-N
SMILES: N[C@@H](CC1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)