accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNOH

CAS RN 172348-63-9

IUPAC Name: tert-butyl N-{[4-(hydroxymethyl)cyclohexyl]methyl}carbamate
Synonyms: tert-butyl N-{[4-(hydroxymethyl)cyclohexyl]methyl}carbamate
Molecular Weight (g/mol): 243.35
Molecular Formula: C13H25NO3
InChi Key: KSXPGASLEFGROT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1CCC(CO)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)