CAS RN 169939-93-9
CAS RN 169939-93-9
IUPAC Name:
18-[(dimethylamino)methyl]-17-oxa-4,14,21-triazahexacyclo[19.6.1.1⁷,¹⁴.0²,⁶.0⁸,¹³.0²²,²⁷]nonacosa-1(28),2(6),7(29),8,10,12,22,24,26-nonaene-3,5-dione hydrochloride
Synonyms:
18-[(dimethylamino)methyl]-17-oxa-4,14,21-triazahexacyclo[19.6.1.1⁷,¹⁴.0²,⁶.0⁸,¹³.0²²,²⁷]nonacosa-1(28),2(6),7(29),8,10,12,22,24,26-nonaene-3,5-dione hydrochloride
Molecular Weight (g/mol):
505.02
Molecular Formula:
C28H29ClN4O3
InChi Key:
NYQIEYDJYFVLPO-UHFFFAOYNA-N
SMILES:
Cl.CN(C)CC1CCN2C=C(C3=CC=CC=C23)C2=C(C(=O)NC2=O)C2=CN(CCO1)C1=CC=CC=C21
1
–
1
of
1
results
1
Ruboxistaurin hydrochloride, MedChemExpress
MedChemExpress Ruboxistaurin (LY333531) hydrochloride is an orally active, selective PKC beta inhibitor (Ki=2 nM). Ruboxistaurin hydrochloride exhibits ATP dependent competitive inhibition of PKC beta I with an IC50 of 4.7 nM. Ruboxistaurin hydrochloride inhibits PKC beta II with an IC50 of 5.9 nM.