accessibility menu, dialog, popup

UserName

CAS RN 16941-92-7

IUPAC Name: ammonium hexachloroiridate
Synonyms: ammonium hexachloroiridate
Molecular Weight (g/mol): 440.99
Molecular Formula: Cl6H8IrN2
InChi Key: VXTVKROMQOXCAJ-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].[Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[Ir]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results