accessibility menu, dialog, popup

UserName

BNNCH3FFFK

CAS RN 1691248-19-7

IUPAC Name: potassium trifluoro[4-(4-methylpiperazin-1-yl)phenyl]boranuide
Synonyms: potassium trifluoro[4-(4-methylpiperazin-1-yl)phenyl]boranuide
Molecular Weight (g/mol): 282.16
Molecular Formula: C11H15BF3KN2
InChi Key: VGVCQMVGOAZLNP-UHFFFAOYSA-N
SMILES: [K+].CN1CCN(CC1)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results