CAS RN 16893-85-9
CAS RN 16893-85-9
IUPAC Name:
disodium hexafluorosilanediuide
Synonyms:
disodium hexafluorosilanediuide
Molecular Weight (g/mol):
188.06
Molecular Formula:
F6Na2Si
InChi Key:
TWGUZEUZLCYTCG-UHFFFAOYSA-N
SMILES:
[Na+].[Na+].F[Si--](F)(F)(F)(F)F
1
–
1
of
1
results
1
Sodium hexafluorosilicate, >99%, Thermo Scientific Chemicals
CAS: 16893-85-9 Molecular Formula: F6Na2Si Molecular Weight (g/mol): 188.06 MDL Number: MFCD00003491 InChI Key: TWGUZEUZLCYTCG-UHFFFAOYSA-N Synonym: sodium hexafluorosilicate,sodium fluosilicate,salufer,sodium silicofluoride,safsan,super prodan,destruxol applex,ortho earwig bait,ortho weevil bait,ens-zem weevil bait PubChem CID: 28127 SMILES: [Na+].[Na+].F[Si--](F)(F)(F)(F)F