accessibility menu, dialog, popup

UserName

CAS RN 167392-57-6

H3COOHNOOHOHOOHOH

CAS RN 167392-57-6

IUPAC Name: 2,3-dihydroxybutanedioic acid; ethyl piperidine-3-carboxylate
Synonyms: (.+-.)-tartaric acid; ethyl piperidine-3-carboxylate
Molecular Weight (g/mol): 307.3
Molecular Formula: C12H21NO8
InChi Key: HHPGQKZOPPDLNH-UHFFFAOYNA-N
SMILES: OC(C(O)C(O)=O)C(O)=O.CCOC(=O)C1CCCNC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results