CAS RN 16712-69-9
CAS RN 16712-69-9
IUPAC Name:
4-ethylbenzene-1-sulfonyl chloride
Synonyms:
4-ethylbenzenesulfonyl chloride
Molecular Weight (g/mol):
204.67
Molecular Formula:
C8H9ClO2S
InChi Key:
LACFLXDRFOQEFZ-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C=C1)S(Cl)(=O)=O
1
–
2
of
2
results
2
4-Ethylbenzenesulfonyl chloride, 97%
CAS: 16712-69-9 Molecular Formula: C8H9ClO2S Molecular Weight (g/mol): 204.668 MDL Number: MFCD00041480 InChI Key: LACFLXDRFOQEFZ-UHFFFAOYSA-N Synonym: 4-ethylbenzene-1-sulfonyl chloride,p-ethylbenzenesulfonyl chloride,4-ethylbenzenesulphonyl chloride,4-ethyl-benzenesulfonyl chloride,ethylbenzenesulfonylchloride,4-ethylbenzenesulfonylchloride,chloro 4-ethylphenyl sulfone,pubchem5133,acmc-209dvn,4-ethylphenylsulfonyl chloride PubChem CID: 570959 IUPAC Name: 4-ethylbenzenesulfonyl chloride SMILES: CCC1=CC=C(C=C1)S(=O)(=O)Cl