accessibility menu, dialog, popup

UserName

CAS RN 166023-29-6

H3CCH3(R)NH3OOCH3CH3H3C

CAS RN 166023-29-6

IUPAC Name: (3R)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Synonyms: (3R)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Molecular Weight (g/mol): 202.32
Molecular Formula: C11H24NO2
InChi Key: WBOPVTYXLFEVGP-SECBINFHSA-O
SMILES: CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)