accessibility menu, dialog, popup

UserName

CAS RN 16582-94-8

H2NOFFFF

CAS RN 16582-94-8

IUPAC Name: 2,3,4,5-tetrafluorobenzamide
Synonyms: 2,3,4,5-tetrafluorobenzamide
Molecular Weight (g/mol): 193.1
Molecular Formula: C7H3F4NO
InChi Key: XMFMIKSWEJRJQE-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results