accessibility menu, dialog, popup

UserName

CAS RN 164334-74-1

FO

CAS RN 164334-74-1

IUPAC Name: 4'-fluoro-[1,1'-biphenyl]-3-carbaldehyde
Synonyms: 4'-fluoro-[1,1'-biphenyl]-3-carbaldehyde
Molecular Weight (g/mol): 200.21
Molecular Formula: C13H9FO
InChi Key: IVYDCJYMOBKHTK-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results