CAS RN 16400-32-1
CAS RN 16400-32-1
IUPAC Name:
1-bromopent-2-yne
Synonyms:
1-bromopent-2-yne
Molecular Weight (g/mol):
147.02
Molecular Formula:
C5H7Br
InChi Key:
VDHGRVFJBGRHMD-UHFFFAOYSA-N
SMILES:
CCC#CCBr
1
–
2
of
2
results
1
1-Bromo-2-pentyne, 97%
CAS: 16400-32-1 Molecular Formula: C5H7Br Molecular Weight (g/mol): 147.02 MDL Number: MFCD00236363 InChI Key: VDHGRVFJBGRHMD-UHFFFAOYSA-N Synonym: 1-bromo-2-pentyne,2-pentynyl bromide,2-pentyne, 1-bromo,1-bromo-pent-2-yne,1-bromo-2-pentine,2-pentyn-1-yl bromide,acmc-1c6g2 PubChem CID: 85399 IUPAC Name: 1-bromopent-2-yne SMILES: CCC#CCBr
2
1-Bromo-2-pentyne, 97%, Thermo Scientific Chemicals
CAS: 16400-32-1 Molecular Formula: C5H7Br Molecular Weight (g/mol): 147.02 MDL Number: MFCD00236363 InChI Key: VDHGRVFJBGRHMD-UHFFFAOYSA-N Synonym: 1-bromo-2-pentyne,2-pentynyl bromide,2-pentyne, 1-bromo,1-bromo-pent-2-yne,1-bromo-2-pentine,2-pentyn-1-yl bromide,acmc-1c6g2 PubChem CID: 85399 IUPAC Name: 1-bromopent-2-yne SMILES: CCC#CCBr