accessibility menu, dialog, popup

UserName

CAS RN 162537-11-3

H3C(S)OHHNCH3CH3OOOCH3

CAS RN 162537-11-3

IUPAC Name: 2-[(methoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Synonyms: 2-[(methoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 189.21
Molecular Formula: C8H15NO4
InChi Key: NWPRXAIYBULIEI-UHFFFAOYNA-N
SMILES: COC(=O)NC(C(O)=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results