CAS RN 162333-02-0
CAS RN 162333-02-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Nitro-4-(trifluoromethyl)benzyl bromide, 97%
CAS: 162333-02-0 Molecular Formula: C8H5BrF3NO3 MDL Number: MFCD03095413