accessibility menu, dialog, popup

UserName

CAS RN 161957-55-7

OOClF

CAS RN 161957-55-7

IUPAC Name: 3-chloro-2-fluorobenzoate
Synonyms: 3-chloro-2-fluorobenzoate
Molecular Weight (g/mol): 173.55
Molecular Formula: C7H3ClFO2
InChi Key: FCSSYEWURMTUSM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(Cl)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)