accessibility menu, dialog, popup

UserName

CAS RN 16097-64-6

FFFClNClN

CAS RN 16097-64-6

IUPAC Name: 2,4-dichloro-6-(trifluoromethyl)pyrimidine
Synonyms: 2,4-dichloro-6-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 216.97
Molecular Formula: C5HCl2F3N2
InChi Key: ZTNFYAJHLPMNSN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results