accessibility menu, dialog, popup

UserName

CAS RN 160194-28-5

CH3SiH3CH3COSiH3CCH3H3C

CAS RN 160194-28-5

IUPAC Name: triethyl({[4-(triethylsilyl)but-3-yn-1-yl]oxy})silane
Synonyms: triethyl({[4-(triethylsilyl)but-3-yn-1-yl]oxy})silane
Molecular Weight (g/mol): 298.62
Molecular Formula: C16H34OSi2
InChi Key: WAQSHASXNKNYSS-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)OCCC#C[Si](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results