accessibility menu, dialog, popup

UserName

OFFFFOHO(E/Z)

CAS RN 1588504-91-9

IUPAC Name: 3-[2-fluoro-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[2-fluoro-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 250.15
Molecular Formula: C10H6F4O3
InChi Key: FGRCNSVSBCJRTF-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=C(F)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results