accessibility menu, dialog, popup

UserName

CAS RN 1578-33-2

CH3SiH3CCH3O

CAS RN 1578-33-2

IUPAC Name: (furan-2-yl)trimethylsilane
Synonyms: furan-2-yltrimethylsilane
Molecular Weight (g/mol): 140.26
Molecular Formula: C7H12OSi
InChi Key: HRWRZEKRKZRDRW-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results