CAS RN 1569-44-4
CAS RN 1569-44-4
IUPAC Name:
3-methylhex-5-en-3-ol
Synonyms:
3-methylhex-5-en-3-ol
Molecular Weight (g/mol):
114.19
Molecular Formula:
C7H14O
InChi Key:
GBARKLJMQRUBKV-UHFFFAOYNA-N
SMILES:
CCC(C)(O)CC=C
1
–
1
of
1
results
1
3-Methyl-5-hexen-3-ol, 98%
CAS: 1569-44-4 Molecular Formula: C7H14O Molecular Weight (g/mol): 114.19 MDL Number: MFCD00021826 InChI Key: GBARKLJMQRUBKV-UHFFFAOYNA-N Synonym: 3-methyl-5-hexen-3-ol,methylaethylallylcarbinol,methylethylallylcarbinol,5-hexen-3-ol, 3-methyl,methylaethylallylcarbinol german,5-hexen-3-ol, 3-methyl-, 3r,acmc-20m3dr,acmc-1bx1u,3-methyl-5-hexene-3-ol PubChem CID: 15288 IUPAC Name: 3-methylhex-5-en-3-ol SMILES: CCC(C)(O)CC=C