accessibility menu, dialog, popup

UserName

H3CONH2FF

CAS RN 155020-51-2

IUPAC Name: 2,3-difluoro-4-methoxyaniline
Synonyms: 2,3-difluoro-4-methoxyaniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: MEETZAMYCFAKKW-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results