CAS RN 1544739-25-4
CAS RN 1544739-25-4
IUPAC Name:
2-[3-(bromomethyl)-4-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-[3-(bromomethyl)-4-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
314.99
Molecular Formula:
C13H17BBrFO2
InChi Key:
BQOMTZAOMQFSJP-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(CBr)=C(F)C=C1
1
–
1
of
1
results
1
3-Bromomethyl-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1544739-25-4 Molecular Formula: C13H17BBrFO2 Molecular Weight (g/mol): 314.989 InChI Key: BQOMTZAOMQFSJP-UHFFFAOYSA-N Synonym: 2-3-bromomethyl-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-3-bromomethyl-4-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane PubChem CID: 75487535 IUPAC Name: 2-[3-(bromomethyl)-4-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)CBr