accessibility menu, dialog, popup

UserName

CAS RN 15378-09-3

IUPAC Name: 1,3-dimethyl 2-[(4-methoxyphenyl)methyl]propanedioate
Synonyms: 1,3-dimethyl 2-[(4-methoxyphenyl)methyl]propanedioate
Molecular Weight (g/mol): 252.27
Molecular Formula: C13H16O5
InChi Key: GXDFOJRQIDXKIH-UHFFFAOYSA-N
SMILES: COC(=O)C(CC1=CC=C(OC)C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results