accessibility menu, dialog, popup

UserName

CAS RN 152152-19-7

IUPAC Name: (2E)-3-(3,4,5-trifluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(3,4,5-trifluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 201.13
Molecular Formula: C9H4F3O2
InChi Key: PHIWFMZBVZFEQJ-OWOJBTEDSA-M
SMILES: [O-]C(=O)\C=C\C1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results