accessibility menu, dialog, popup

UserName

CAS RN 151-63-3

NH2NOHSOHOO

CAS RN 151-63-3

IUPAC Name: 2-aminoacetonitrile; sulfuric acid
Synonyms: aminoacetonitrile; sulfuric acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C2H6N2O4S
InChi Key: GTGIXCPOLMWQTC-UHFFFAOYSA-N
SMILES: NCC#N.OS(O)(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results