CAS RN 150349-36-3
CAS RN 150349-36-3
IUPAC Name:
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Synonyms:
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Molecular Weight (g/mol):
188.27
Molecular Formula:
C9H20N2O2
InChi Key:
PNQYAMWGTGWJDW-UHFFFAOYSA-N
SMILES:
CN(CCCN)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals
CAS: 150349-36-3 Molecular Formula: C9H20N2O2 Molecular Weight (g/mol): 188.271 MDL Number: MFCD01631189 InChI Key: PNQYAMWGTGWJDW-UHFFFAOYSA-N Synonym: n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester,tert-butyl n-3-aminopropyl-n-methylcarbamate,tert-butyl 3-aminopropyl methyl carbamate,3-n-boc-n-methylamino propylamine,n-boc-n-methyl-1,3-diaminopropane,n-3-amino-propyl-n-methylcarbamic acid t-butyl ester,acid tert-butyl ester,3-n-tert-butoxycarbonyl-n-methylamino propylamine,tert-butyl n-3-aminopropyl-n-methyl carbamate PubChem CID: 2756543 IUPAC Name: tert-butyl N-(3-aminopropyl)-N-methylcarbamate SMILES: CC(C)(C)OC(=O)N(C)CCCN