accessibility menu, dialog, popup

UserName

CAS RN 148637-74-5

SNClOCl

CAS RN 148637-74-5

IUPAC Name: 2-chloro-1,3-thiazole-5-carbonyl chloride
Synonyms: 2-chloro-1,3-thiazole-5-carbonyl chloride
Molecular Weight (g/mol): 182.02
Molecular Formula: C4HCl2NOS
InChi Key: GZSJPOBGZMUSPV-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CN=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results