accessibility menu, dialog, popup

UserName

CAS RN 145549-76-4

CH3H3CCH3OOO

CAS RN 145549-76-4

IUPAC Name: tert-butyl 3-oxocyclobutane-1-carboxylate
Synonyms: tert-butyl 3-oxocyclobutane-1-carboxylate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: JINYZTGTQXDUQR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1CC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results