CAS RN 14529-53-4
CAS RN 14529-53-4
IUPAC Name:
2-ethoxypyridine
Synonyms:
2-ethoxypyridine
Molecular Weight (g/mol):
123.16
Molecular Formula:
C7H9NO
InChi Key:
LISKAOIANGDBTB-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=N1
1
–
2
of
2
results
1
2-Ethoxypyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Ethoxypyridine, 94%, Thermo Scientific™
CAS: 14529-53-4 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.16 MDL Number: MFCD00023464 InChI Key: LISKAOIANGDBTB-UHFFFAOYSA-N Synonym: pyridine, 2-ethoxy,2-ethoxy pyridine,2-ethoxy-pyridine,zlchem 304,pubchem1126,acmc-209cun,ethyl 2-pyridinyl ether,ksc497c7d PubChem CID: 84499 IUPAC Name: 2-ethoxypyridine SMILES: CCOC1=CC=CC=N1