accessibility menu, dialog, popup

UserName

CAS RN 144100-07-2

FBrN

CAS RN 144100-07-2

IUPAC Name: 2-bromo-6-fluoropyridine
Synonyms: 2-bromo-6-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: ZIDIKYIZXMYHAW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results