accessibility menu, dialog, popup

UserName

CAS RN 1439402-07-9

IUPAC Name: potassium [4-(ethoxycarbonyl)phenyl]trifluoroboranuide
Synonyms: potassium [4-(ethoxycarbonyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 256.07
Molecular Formula: C9H9BF3KO2
InChi Key: RRHVYQWNJHNUEQ-UHFFFAOYSA-N
SMILES: [K+].CCOC(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results