CAS RN 1433953-83-3
CAS RN 1433953-83-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SAR131675, MedChemExpress
MedChemExpress SAR131675 is a potent and selective VEGFR3 inhibitor with an IC50 of 23 nM.