accessibility menu, dialog, popup

UserName

CAS RN 1431329-75-7

CH3ONH2FFFOCH3

CAS RN 1431329-75-7

IUPAC Name: 4-methoxy-3-methyl-5-(trifluoromethyl)benzamide
Synonyms: 4-methoxy-3-methyl-5-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 233.19
Molecular Formula: C10H10F3NO2
InChi Key: XXEJFZMPVFRXHR-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1C(F)(F)F)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results