CAS RN 1418741-86-2
CAS RN 1418741-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
UNC1079, MedChemExpress
MedChemExpress UNC1079 is the piperidine analog of UNC1021, as a structurally similar but significantly less potent inhibitor for use as a negative control in cellular studies.