accessibility menu, dialog, popup

UserName

CAS RN 1397238-97-9

IUPAC Name: N-(2,4,6-tribromophenyl)benzenesulfonamide
Synonyms: N-(2,4,6-tribromophenyl)benzenesulfonamide
Molecular Weight (g/mol): 469.97
Molecular Formula: C12H8Br3NO2S
InChi Key: FDGMNRHQLZRBGJ-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(NS(=O)(=O)C2=CC=CC=C2)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results