CAS RN 1391194-67-4
CAS RN 1391194-67-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2R)-Octyl-α-hydroxyglutarate, MedChemExpress
MedChemExpress (2R)-Octyl-α-hydroxyglutarate ((2R)-Octyl-2-HG) is a modified form of D-isomer 2-Hydroxyglutarate.