accessibility menu, dialog, popup

UserName

CAS RN 1387445-51-3

IUPAC Name: 4-aminocyclohexane-1-carbonitrile hydrochloride
Synonyms: 4-aminocyclohexane-1-carbonitrile hydrochloride
Molecular Weight (g/mol): 160.65
Molecular Formula: C7H13ClN2
InChi Key: BTLVSLNZSMIPCW-UHFFFAOYSA-N
SMILES: Cl.NC1CCC(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results